Assay Design



Gene Transcripts

Gene Symbol
TTC33
Entrez Gene ID
531278
Gene Name
tetratricopeptide repeat domain 33
Gene Aliases
-
Chromosome Location
Chr.20: 33708356 - 33737405 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192705.1 NP_001179634.1 4 - 5 478 73

View Full Product Details