Assay Design



Gene Transcripts

Gene Symbol
GLA
Entrez Gene ID
532742
Gene Name
galactosidase alpha
Gene Aliases
-
Chromosome Location
Chr.X: 50803846 - 50814062 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192736.1 NP_001179665.1 1 - 2 299 66

View Full Product Details