Assay Design



Gene Transcripts

Gene Symbol
ACER2
Entrez Gene ID
540732
Gene Name
alkaline ceramidase 2
Gene Aliases
ASAH3L
Chromosome Location
Chr.8: 24859498 - 24891823 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192775.1 NP_001179704.1 4 - 5 508 71

View Full Product Details