Assay Design



Gene Transcripts

Gene Symbol
SAMD10
Entrez Gene ID
540999
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
-
Chromosome Location
Chr.13: 53873001 - 53877879 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192805.1 NP_001179734.1 2 - 3 271 104

View Full Product Details