Assay Design



Gene Transcripts

Gene Symbol
SLC16A10
Entrez Gene ID
541240
Gene Name
solute carrier family 16 member 10
Gene Aliases
MCT10, TAT1
Chromosome Location
Chr.9: 39168422 - 39280816 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192847.1 NP_001179776.1 3 - 4 941 102
GenBank mRNA AB435507.1 - 2 - 3 534 102

View Full Product Details