Assay Design



Gene Transcripts

Gene Symbol
SMAD7
Entrez Gene ID
535916
Gene Name
SMAD family member 7
Gene Aliases
-
Chromosome Location
Chr.24: 48598698 - 48627436 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192865.1 NP_001179794.1 1 - 2 614 95

View Full Product Details