Assay Design



Gene Transcripts

Gene Symbol
OVOS2
Entrez Gene ID
508026
Gene Name
ovostatin 2
Gene Aliases
OVOS
Chromosome Location
Chr.5: 100679985 - 100732010 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164024.1 NP_001157496.1 30 - 31 4131 105
GenBank mRNA BC111679.1 - 1 - 2 94 105

View Full Product Details