Assay Design



Gene Transcripts

Gene Symbol
BCL2
Entrez Gene ID
281020
Gene Name
BCL2 apoptosis regulator
Gene Aliases
-
Chromosome Location
Chr.24: 61396513 - 61589114 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166486.1 NP_001159958.1 1 - 2 554 72
GenBank mRNA U92434.1 - 1 - 2 554 72

View Full Product Details