Assay Design



Gene Transcripts

Gene Symbol
PRPH2
Entrez Gene ID
280907
Gene Name
peripherin 2
Gene Aliases
RDS
Chromosome Location
Chr.23: 16367966 - 16382111 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166487.1 NP_001159959.1 1 - 2 587 70
GenBank mRNA J02884.1 - 1 - 2 569 70

View Full Product Details