Assay Design



Gene Transcripts

Gene Symbol
TCHHL1
Entrez Gene ID
518825
Gene Name
trichohyalin like 1
Gene Aliases
-
Chromosome Location
Chr.3: 18642146 - 18648444 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206595.1 NP_001193524.1 1 - 2 162 92
GenBank mRNA BC149208.1 - 1 - 2 162 92

View Full Product Details