Assay Design



Gene Transcripts

Gene Symbol
CCDC69
Entrez Gene ID
529878
Gene Name
coiled-coil domain containing 69
Gene Aliases
-
Chromosome Location
Chr.7: 62434491 - 62469922 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206624.1 NP_001193553.1 2 - 3 234 60
GenBank mRNA BC148945.1 - 2 - 3 234 60

View Full Product Details