Assay Design



Gene Transcripts

Gene Symbol
FGF8
Entrez Gene ID
326284
Gene Name
fibroblast growth factor 8
Gene Aliases
-
Chromosome Location
Chr.26: 22503903 - 22509709 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206678.1 NP_001193607.1 3 - 4 350 89
GenBank mRNA CN433133.1 - 3 - 4 354 89

View Full Product Details