Assay Design



Gene Transcripts

Gene Symbol
GRIFIN
Entrez Gene ID
528362
Gene Name
galectin-related inter-fiber protein
Gene Aliases
-
Chromosome Location
Chr.25: 40799380 - 40801701 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206700.1 NP_001193629.1 4 - 5 423 116
GenBank mRNA EG707677.1 - 4 - 5 426 116

View Full Product Details