Assay Design



Gene Transcripts

Gene Symbol
P2RY2
Entrez Gene ID
282638
Gene Name
purinergic receptor P2Y2
Gene Aliases
-
Chromosome Location
Chr.15: 52709682 - 52730947 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166525.1 NP_001159997.1 1 - 2 167 80

View Full Product Details