Assay Design



Gene Transcripts

Gene Symbol
UPK3BL1
Entrez Gene ID
617471
Gene Name
uroplakin 3B-like
Gene Aliases
UPK3BL, UPLP
Chromosome Location
Chr.25: 34519527 - 34524688 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168011.2 NP_001161483.2 3 - 4 551 69
GenBank mRNA BC149698.1 - 3 - 4 534 69

View Full Product Details