Assay Design



Gene Transcripts

Gene Symbol
SLC8B1
Entrez Gene ID
508887
Gene Name
solute carrier family 8 member B1
Gene Aliases
SLC24A6
Chromosome Location
Chr.17: 61286699 - 61316746 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001177596.1 NP_001171067.1 1 - 2 131 71
GenBank mRNA DY054190.1 - 1 - 2 146 71

View Full Product Details