Assay Design



Gene Transcripts

Gene Symbol
INS
Entrez Gene ID
280829
Gene Name
insulin
Gene Aliases
-
Chromosome Location
Chr.29: 49392917 - 49394536 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001185126.1 NP_001172055.1 1 - 2 47 65

View Full Product Details