Assay Design



Gene Transcripts

Gene Symbol
LHFPL1
Entrez Gene ID
781310
Gene Name
LHFPL tetraspan subfamily member 1
Gene Aliases
-
Chromosome Location
Chr.X: 61079935 - 61150832 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191127.1 NP_001178056.1 2 - 3 469 90

View Full Product Details