Assay Design



Gene Transcripts

Gene Symbol
VAX1
Entrez Gene ID
782056
Gene Name
ventral anterior homeobox 1
Gene Aliases
-
Chromosome Location
Chr.26: 37467830 - 37472057 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191142.1 NP_001178071.1 1 - 2 241 90

View Full Product Details