Assay Design



Gene Transcripts

Gene Symbol
CHSY1
Entrez Gene ID
281690
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
-
Chromosome Location
Chr.21: 5285464 - 5375159 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191157.1 NP_001178086.1 2 - 3 823 77
GenBank mRNA AF440749.1 - 1 - 2 358 77

View Full Product Details