Assay Design



Gene Transcripts

Gene Symbol
CEP131
Entrez Gene ID
514944
Gene Name
centrosomal protein 131
Gene Aliases
AZI1
Chromosome Location
Chr.19: 51500742 - 51517016 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191187.3 NP_001178116.3 1 - 2 174 93

View Full Product Details