Assay Design



Gene Transcripts

Gene Symbol
GGT5
Entrez Gene ID
787326
Gene Name
gamma-glutamyltransferase 5
Gene Aliases
GGTLA1
Chromosome Location
Chr.17: 71443755 - 71455166 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191219.3 NP_001178148.1 7 - 8 1039 102

View Full Product Details