Assay Design



Gene Transcripts

Gene Symbol
IGLON5
Entrez Gene ID
786369
Gene Name
IgLON family member 5
Gene Aliases
-
Chromosome Location
Chr.18: 57390098 - 57404602 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206875.1 NP_001193804.1 6 - 7 766 77
GenBank mRNA EE351204.1 - 4 - 5 418 77
EH200051.1 - 6 - 7 743 77

View Full Product Details