Assay Design



Gene Transcripts

Gene Symbol
SLC16A2
Entrez Gene ID
535872
Gene Name
solute carrier family 16 member 2
Gene Aliases
MCT8
Chromosome Location
Chr.X: 76549545 - 76681857 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001206939.1 NP_001193868.1 1 - 2 624 102
GenBank mRNA DV924764.1 - 1 - 2 506 102

View Full Product Details