Assay Design



Gene Transcripts

Gene Symbol
ATP6AP1L
Entrez Gene ID
510390
Gene Name
ATPase H+ transporting accessory protein 1 like
Gene Aliases
-
Chromosome Location
Chr.7: 82237768 - 82260744 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207027.1 NP_001193956.1 2 - 3 212 92
GenBank mRNA EE894106.1 - 2 - 3 187 92

View Full Product Details