Assay Design



Gene Transcripts

Gene Symbol
ANKRD34B
Entrez Gene ID
529927
Gene Name
ankyrin repeat domain 34B
Gene Aliases
-
Chromosome Location
Chr.7: 80676426 - 80694011 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001207060.1 NP_001193989.1 2 - 3 215 81
GenBank mRNA BC122789.1 - 2 - 3 216 81

View Full Product Details