Assay Design



Gene Transcripts

Gene Symbol
KNCN
Entrez Gene ID
100139285
Gene Name
kinocilin
Gene Aliases
-
Chromosome Location
Chr.3: 99576630 - 99584715 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191306.1 NP_001178235.1 2 - 3 224 83

View Full Product Details