Assay Design



Gene Transcripts

Gene Symbol
WDR86
Entrez Gene ID
100140069
Gene Name
WD repeat domain 86
Gene Aliases
-
Chromosome Location
Chr.4: 113914551 - 113956869 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191339.2 NP_001178268.2 6 - 7 1182 60
GenBank mRNA BC149915.1 - 5 - 6 831 60

View Full Product Details