Assay Design



Gene Transcripts

Gene Symbol
GUCY1A2
Entrez Gene ID
613600
Gene Name
guanylate cyclase 1 soluble subunit alpha 2
Gene Aliases
-
Chromosome Location
Chr.15: 16344389 - 16845470 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191341.1 NP_001178270.1 5 - 6 1686 78

View Full Product Details