Assay Design



Gene Transcripts

Gene Symbol
NR3C2
Entrez Gene ID
537614
Gene Name
nuclear receptor subfamily 3 group C member 2
Gene Aliases
-
Chromosome Location
Chr.17: 9581384 - 10021369 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191349.2 NP_001178278.1 6 - 7 2643 80

View Full Product Details