Assay Design



Gene Transcripts

Gene Symbol
ESRRA
Entrez Gene ID
507834
Gene Name
estrogen related receptor alpha
Gene Aliases
-
Chromosome Location
Chr.29: 42603705 - 42611807 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191373.2 NP_001178302.1 4 - 5 738 77

View Full Product Details