Assay Design



Gene Transcripts

Gene Symbol
LRRIQ4
Entrez Gene ID
784301
Gene Name
leucine rich repeats and IQ motif containing 4
Gene Aliases
-
Chromosome Location
Chr.1: 97542149 - 97557519 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191385.1 NP_001178314.1 5 - 6 1665 104

View Full Product Details