Assay Design



Gene Transcripts

Gene Symbol
TRIM8
Entrez Gene ID
782444
Gene Name
tripartite motif containing 8
Gene Aliases
-
Chromosome Location
Chr.26: 23265725 - 23279452 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191476.2 NP_001178405.1 2 - 3 943 73
GenBank mRNA EV635295.1 - 1 - 2 101 73

View Full Product Details