Assay Design



Gene Transcripts

Gene Symbol
GSDMB
Entrez Gene ID
509296
Gene Name
gasdermin B
Gene Aliases
GSDML
Chromosome Location
Chr.19: 40242818 - 40256060 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244217.1 NP_001231146.1 1 - 2 68 115
GenBank mRNA BC149305.1 - 1 - 2 69 115

View Full Product Details