Assay Design
Gene Transcripts
- Gene Symbol
- CYLC1
- Entrez Gene ID
-
281737
- Gene Name
- cylicin 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 68899563 - 68943644 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174302.3 |
NP_776727.1 |
2 - 3 |
91 |
91 |
GenBank mRNA |
Z22779.1 |
- |
2 - 3 |
92 |
91 |
View Full Product Details