Assay Design



Gene Transcripts

Gene Symbol
CHST6
Entrez Gene ID
538464
Gene Name
carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6
Gene Aliases
-
Chromosome Location
Chr.18: 2822521 - 2841277 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192022.1 NP_001178951.1 1 - 2 29 84

View Full Product Details