Assay Design



Gene Transcripts

Gene Symbol
SLC25A42
Entrez Gene ID
504608
Gene Name
solute carrier family 25 member 42
Gene Aliases
-
Chromosome Location
Chr.7: 4121449 - 4162586 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192032.1 NP_001178961.1 3 - 4 213 86

View Full Product Details