Assay Design



Gene Transcripts

Gene Symbol
SLC7A4
Entrez Gene ID
504923
Gene Name
solute carrier family 7 member 4
Gene Aliases
-
Chromosome Location
Chr.17: 72373077 - 72376852 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192042.1 NP_001178971.1 2 - 3 1718 67

View Full Product Details