Assay Design



Gene Transcripts

Gene Symbol
FOXJ1
Entrez Gene ID
505891
Gene Name
forkhead box J1
Gene Aliases
-
Chromosome Location
Chr.19: 55540965 - 55545081 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192076.1 NP_001179005.1 1 - 2 498 78

View Full Product Details