Assay Design



Gene Transcripts

Gene Symbol
MYBPHL
Entrez Gene ID
538875
Gene Name
myosin binding protein H like
Gene Aliases
-
Chromosome Location
Chr.3: 34107381 - 34122171 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192119.1 NP_001179048.1 4 - 5 567 53

View Full Product Details