Assay Design



Gene Transcripts

Gene Symbol
TBX21
Entrez Gene ID
538933
Gene Name
T-box transcription factor 21
Gene Aliases
-
Chromosome Location
Chr.19: 38715293 - 38727078 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192140.1 NP_001179069.1 2 - 3 645 86

View Full Product Details