Assay Design



Gene Transcripts

Gene Symbol
PGAM5
Entrez Gene ID
508608
Gene Name
PGAM family member 5, mitochondrial serine/threonine protein phosphatase
Gene Aliases
-
Chromosome Location
Chr.17: 44502890 - 44510533 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192191.1 NP_001179120.1 2 - 3 420 75
GenBank mRNA BC149319.1 - 2 - 3 413 75

View Full Product Details