Assay Design



Gene Transcripts

Gene Symbol
DNAJC21
Entrez Gene ID
509302
Gene Name
DnaJ heat shock protein family (Hsp40) member C21
Gene Aliases
-
Chromosome Location
Chr.20: 39187820 - 39214684 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192218.3 NP_001179147.1 4 - 5 589 96
GenBank mRNA BC122750.1 - 4 - 5 596 96

View Full Product Details