Assay Design



Gene Transcripts

Gene Symbol
THUMPD2
Entrez Gene ID
509460
Gene Name
THUMP domain containing 2
Gene Aliases
-
Chromosome Location
Chr.11: 22040726 - 22080810 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192222.3 NP_001179151.2 7 - 8 968 98
GenBank mRNA BC149576.1 - 7 - 8 671 98

View Full Product Details