Assay Design



Gene Transcripts

Gene Symbol
RTKN2
Entrez Gene ID
539797
Gene Name
rhotekin 2
Gene Aliases
-
Chromosome Location
Chr.28: 18280636 - 18472006 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192310.2 NP_001179239.2 11 - 12 1394 115

View Full Product Details