Assay Design



Gene Transcripts

Gene Symbol
PRKAB2
Entrez Gene ID
512665
Gene Name
protein kinase AMP-activated non-catalytic subunit beta 2
Gene Aliases
-
Chromosome Location
Chr.3: 22529653 - 22545871 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192328.1 NP_001179257.1 3 - 4 447 65

View Full Product Details