Assay Design



Gene Transcripts

Gene Symbol
TFAP2D
Entrez Gene ID
539910
Gene Name
transcription factor AP-2 delta
Gene Aliases
-
Chromosome Location
Chr.23: 23277603 - 23337345 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192329.1 NP_001179258.1 3 - 4 1116 76

View Full Product Details