Assay Design



Gene Transcripts

Gene Symbol
ARL11
Entrez Gene ID
513113
Gene Name
ADP ribosylation factor like GTPase 11
Gene Aliases
-
Chromosome Location
Chr.12: 19179628 - 19203345 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192342.1 NP_001179271.1 2 - 3 144 88

View Full Product Details