Assay Design



Gene Transcripts

Gene Symbol
ARHGAP35
Entrez Gene ID
540310
Gene Name
Rho GTPase activating protein 35
Gene Aliases
GRLF1
Chromosome Location
Chr.18: 53947317 - 54063061 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192398.1 NP_001179327.1 3 - 4 3910 96

View Full Product Details