Assay Design



Gene Transcripts

Gene Symbol
CHP2
Entrez Gene ID
515280
Gene Name
calcineurin like EF-hand protein 2
Gene Aliases
-
Chromosome Location
Chr.25: 21402614 - 21406033 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192405.2 NP_001179334.1 1 - 2 278 76

View Full Product Details